Learn More
Pacer Global LLC Saint-Gobain - SaniTech-C Tubing, 0.15625'' ID, 0.40625'' OD, 0.125'' Wall, 50 Foot Coil, 1/EA - STHT-C-156-4F
SDP
Catalog No. 5000332248
Encompass_Preferred
Saint-Gobain - SaniTech-C Tubing, 0.15625" ID, 0.40625" OD, 0.125" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32248 Supplier Pacer Global LLC Supplier No. MLSSTHTC1564F
Saint-Gobain - SaniTech-C Tubing, 0.15625" ID, 0.40625" OD, 0.125" Wall, 50 Foot Coil, 1/EA