Learn More
Pacer Global LLC Saint-Gobain - SaniTech-C Tubing, 0.1875'' ID, 0.3125'' OD, 0.0625'' Wall, 25 Foot Coil, 1/EA - STHT-C-187-2
SDP
Catalog No. 5000332282
Encompass_Preferred
Saint-Gobain - SaniTech-C Tubing, 0.1875" ID, 0.3125" OD, 0.0625" Wall, 25 Foot Coil, 1/EA
Catalog No. 50-003-32282 Supplier Pacer Global LLC Supplier No. MLSSTHTC1872
Saint-Gobain - SaniTech-C Tubing, 0.1875" ID, 0.3125" OD, 0.0625" Wall, 25 Foot Coil, 1/EA