Learn More
Pacer Global LLC Saint-Gobain - SaniTech-C Tubing, 0.5'' ID, 0.875'' OD, 0.1875'' Wall, 100 Foot Coil, 1/EA - STHT-C-500-5H
SDP
Catalog No. 5000332321
Encompass_Preferred
Saint-Gobain - SaniTech-C Tubing, 0.5" ID, 0.875" OD, 0.1875" Wall, 100 Foot Coil, 1/EA
Catalog No. 50-003-32321 Supplier Pacer Global LLC Supplier No. MLSSTHTC5005H
Saint-Gobain - SaniTech-C Tubing, 0.5" ID, 0.875" OD, 0.1875" Wall, 100 Foot Coil, 1/EA