Learn More
Pacer Global LLC Saint-Gobain - SaniTech-C Tubing, 0.625'' ID, 0.875'' OD, 0.125'' Wall, 50 Foot Coil, 1/EA - STHT-C-625-4F
SDP
Catalog No. 5000332271
Encompass_Preferred
Saint-Gobain - SaniTech-C Tubing, 0.625" ID, 0.875" OD, 0.125" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32271 Supplier Pacer Global LLC Supplier No. MLSSTHTC6254F
Saint-Gobain - SaniTech-C Tubing, 0.625" ID, 0.875" OD, 0.125" Wall, 50 Foot Coil, 1/EA