Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-65 Tubing, 0.3125'' ID, 0.4375'' OD, 0.0625'' Wall, 50 Foot Coil, 1/EA - Ultra65-312-2F
SDP
Catalog No. 5000332450
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-65 Tubing, 0.3125" ID, 0.4375" OD, 0.0625" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32450 Supplier Pacer Global LLC Supplier No. MLSULTRA653122F
Saint-Gobain - SaniTech Ultra-65 Tubing, 0.3125" ID, 0.4375" OD, 0.0625" Wall, 50 Foot Coil, 1/EA