Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-65 Tubing, 0.3125'' ID, 0.5'' OD, 0.09375'' Wall, 100 Foot Coil, 1/EA - Ultra65-312-3H
SDP
Catalog No. 5000332501
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-65 Tubing, 0.3125" ID, 0.5" OD, 0.09375" Wall, 100 Foot Coil, 1/EA
Catalog No. 50-003-32501 Supplier Pacer Global LLC Supplier No. MLSULTRA653123H
Saint-Gobain - SaniTech Ultra-65 Tubing, 0.3125" ID, 0.5" OD, 0.09375" Wall, 100 Foot Coil, 1/EA