Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-65 Tubing, 0.375'' ID, 0.625'' OD, 0.125'' Wall, 50 Foot Coil, 1/EA - Ultra65-375-4F
SDP
Catalog No. 5000332201
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-65 Tubing, 0.375" ID, 0.625" OD, 0.125" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32201 Supplier Pacer Global LLC Supplier No. MLSULTRA653754F
Saint-Gobain - SaniTech Ultra-65 Tubing, 0.375" ID, 0.625" OD, 0.125" Wall, 50 Foot Coil, 1/EA