Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-C Tubing, 0.04'' ID, 0.086'' OD, 0.023'' Wall, 50 Foot Coil, 1/EA - ULTRA-C-040-0F
SDP
Catalog No. 5000332381
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-C Tubing, 0.04" ID, 0.086" OD, 0.023" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32381 Supplier Pacer Global LLC Supplier No. MLSULTRAC0400F
Saint-Gobain - SaniTech Ultra-C Tubing, 0.04" ID, 0.086" OD, 0.023" Wall, 50 Foot Coil, 1/EA