Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-C Tubing, 0.0625'' ID, 0.25'' OD, 0.09375'' Wall, 50 Foot Coil, 1/EA - ULTRA-C-062-3F
SDP
Catalog No. 5000332187
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-C Tubing, 0.0625" ID, 0.25" OD, 0.09375" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32187 Supplier Pacer Global LLC Supplier No. MLSULTRAC0623F
Saint-Gobain - SaniTech Ultra-C Tubing, 0.0625" ID, 0.25" OD, 0.09375" Wall, 50 Foot Coil, 1/EA