Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-C Tubing, 0.09375'' ID, 0.28125'' OD, 0.09375'' Wall, 50 Foot Coil, 1/EA - ULTRA-C-093-3F
SDP
Catalog No. 5000332327
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-C Tubing, 0.09375" ID, 0.28125" OD, 0.09375" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32327 Supplier Pacer Global LLC Supplier No. MLSULTRAC0933F
Saint-Gobain - SaniTech Ultra-C Tubing, 0.09375" ID, 0.28125" OD, 0.09375" Wall, 50 Foot Coil, 1/EA