Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-C Tubing, 0.125'' ID, 0.25'' OD, 0.0625'' Wall, 100 Foot Coil, 1/EA - ULTRA-C-125-2H
SDP
Catalog No. 5000332434
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-C Tubing, 0.125" ID, 0.25" OD, 0.0625" Wall, 100 Foot Coil, 1/EA
Catalog No. 50-003-32434 Supplier Pacer Global LLC Supplier No. MLSULTRAC1252H
Saint-Gobain - SaniTech Ultra-C Tubing, 0.125" ID, 0.25" OD, 0.0625" Wall, 100 Foot Coil, 1/EA