Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-C Tubing, 0.125'' ID, 0.3125'' OD, 0.09375'' Wall, 50 Foot Coil, 1/EA - ULTRA-C-125-3F
SDP
Catalog No. 5000332325
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-C Tubing, 0.125" ID, 0.3125" OD, 0.09375" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32325 Supplier Pacer Global LLC Supplier No. MLSULTRAC1253F
Saint-Gobain - SaniTech Ultra-C Tubing, 0.125" ID, 0.3125" OD, 0.09375" Wall, 50 Foot Coil, 1/EA