Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-C Tubing, 0.25'' ID, 0.375'' OD, 0.0625'' Wall, 25 Foot Coil, 1/EA - ULTRA-C-250-2
SDP
Catalog No. 5000332423
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-C Tubing, 0.25" ID, 0.375" OD, 0.0625" Wall, 25 Foot Coil, 1/EA
Catalog No. 50-003-32423 Supplier Pacer Global LLC Supplier No. MLSULTRAC2502
Saint-Gobain - SaniTech Ultra-C Tubing, 0.25" ID, 0.375" OD, 0.0625" Wall, 25 Foot Coil, 1/EA