Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-C Tubing, 0.3125'' ID, 0.5'' OD, 0.09375'' Wall, 25 Foot Coil, 1/EA - ULTRA-C-312-3
SDP
Catalog No. 5000332490
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-C Tubing, 0.3125" ID, 0.5" OD, 0.09375" Wall, 25 Foot Coil, 1/EA
Catalog No. 50-003-32490 Supplier Pacer Global LLC Supplier No. MLSULTRAC3123
Saint-Gobain - SaniTech Ultra-C Tubing, 0.3125" ID, 0.5" OD, 0.09375" Wall, 25 Foot Coil, 1/EA