Learn More
Pacer Global LLC Saint-Gobain - SaniTech Ultra-C Tubing, 0.75'' ID, 1.25'' OD, 0.25'' Wall, 50 Foot Coil, 1/EA - ULTRA-C-750-6F
SDP
Catalog No. 5000332446
Encompass_Preferred
Saint-Gobain - SaniTech Ultra-C Tubing, 0.75" ID, 1.25" OD, 0.25" Wall, 50 Foot Coil, 1/EA
Catalog No. 50-003-32446 Supplier Pacer Global LLC Supplier No. MLSULTRAC7506F
Saint-Gobain - SaniTech Ultra-C Tubing, 0.75" ID, 1.25" OD, 0.25" Wall, 50 Foot Coil, 1/EA