Learn More
Accuflow Systems Saint-Gobain SIB Retainer Connector, Polypropylene, 1/2 in, 250 psi, 250/Pk.
Catalog No. NC3936001
Encompass_Preferred
The Pure-Fit SIB (Smooth Inner Bore) hose-barb connector is designed to provide a seamless transition between tubing and fittings within the fluid path. This design eliminates entrapment and leak points often associated with traditional assembly systems, preventing voids or dead spaces that can lead to fluid stagnation or bacterial growth.
  • Eliminates entrapment and leak points common in traditional fitting systems.
  • Polypropylene material is gamma stable.
  • Materials are completely animal-free and BPA-free.
  • Manufactured and packaged in a certified clean room.
  • Meets USP <88> Class VI, and/or USP <87>, and/or ISO 10993-5 requirements.
  • Smooth inner bore for enhanced flow.
  • Durable construction for reliable performance.
Catalog No. NC3936001 Supplier Accuflow Systems Supplier No. PF135500PPAF
The Pure-Fit SIB (Smooth Inner Bore) hose-barb connector is designed to provide a seamless transition between tubing and fittings within the fluid path. This design eliminates entrapment and leak points often associated with traditional assembly systems, preventing voids or dead spaces that can lead to fluid stagnation or bacterial growth.
  • Eliminates entrapment and leak points common in traditional fitting systems.
  • Polypropylene material is gamma stable.
  • Materials are completely animal-free and BPA-free.
  • Manufactured and packaged in a certified clean room.
  • Meets USP <88> Class VI, and/or USP <87>, and/or ISO 10993-5 requirements.
  • Smooth inner bore for enhanced flow.
  • Durable construction for reliable performance.