Saint-Gobain Tygon S3 B-44-4X tubing is a clear, flexible, unreinforced tube engineered for food and dairy fluid-transfer applications. It provides chemical resistance to common sanitizers and alkaline cleaners, and is formulated without BPA or phthalates.
- Clear, flexible, unreinforced tubing for food and dairy transfer.
- Inner diameter 3/4 in (19.05 mm), outer diameter 1 in (25.40 mm), wall thickness 1/8 in (3.18 mm).
- Package length 50 ft (15.24 m).
- Material: PVC construction (listed by distributors).
- Maximum recommended operating temperature 165°F (74°C); brittleness by impact -32°F (-36°C).
- Maximum working pressure 25 psi (1.7 bar) at 73°F (23°C).
- Contains no BPA or phthalates; resists commonly used sanitizers and harsh alkaline cleaners.
- Common applications: aseptic filling, dairy processing, condiment dispensing, vitamin and flavor concentrate systems, soft-serve dispensing.