Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Plasticizer Free, 3/16 in ID, 50 ft
Catalog No. NC3852764
Encompass_Preferred
Tygon 2475 high-purity, plasticizer-free tubing is a crystal-clear thermoplastic designed for medical, diagnostic, and laboratory fluid transfer where low extractables, chemical resistance, and biocompatibility are required. It resists aggressive acids, bases, and solvents, and offers lower gas permeation than silicone to help preserve fluid integrity.
  • Plasticizer-free formulation reduces extractables and contamination risk.
  • Low gas permeation compared with silicone to minimize moisture loss and oxidation.
  • Smooth, transparent inner bore for reduced particle entrapment and visual inspection.
  • Meets USP Class VI biocompatibility standards.
  • Dimensions: 3/16 in ID, 5/16 in OD, 1/16 in wall.
  • Package length: 50 ft (15.2 m) for long runs and reduced joins.
  • Suitable for diagnostic reagent delivery, medical dispensing, and cell and tissue culture transfer.
Catalog No. NC3852764 Supplier Accuflow Systems Supplier No. ACG00012
Tygon 2475 high-purity, plasticizer-free tubing is a crystal-clear thermoplastic designed for medical, diagnostic, and laboratory fluid transfer where low extractables, chemical resistance, and biocompatibility are required. It resists aggressive acids, bases, and solvents, and offers lower gas permeation than silicone to help preserve fluid integrity.
  • Plasticizer-free formulation reduces extractables and contamination risk.
  • Low gas permeation compared with silicone to minimize moisture loss and oxidation.
  • Smooth, transparent inner bore for reduced particle entrapment and visual inspection.
  • Meets USP Class VI biocompatibility standards.
  • Dimensions: 3/16 in ID, 5/16 in OD, 1/16 in wall.
  • Package length: 50 ft (15.2 m) for long runs and reduced joins.
  • Suitable for diagnostic reagent delivery, medical dispensing, and cell and tissue culture transfer.