Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Silicone, Food and Beverage, 1/4 in ID
Catalog No. NC3877233
Encompass_Preferred
Tygon SPT-3350 platinum-cured silicone tubing is engineered for sanitary food and beverage transfer. It features an ultra-smooth inner bore to minimize particle entrapment, low extractables to preserve fluid integrity, and a wide operating temperature range suitable for processing and sterilization.
  • Platinum-cured silicone.
  • Ultra-smooth inner bore reduces particle entrapment.
  • FDA 21 CFR 175.300 compliant.
  • NSF 51 compliant where applicable.
  • Available in sizes including 1/4 in ID x 7/16 in OD, 3/32 in wall, 50 ft coil.
  • Operating temperature range -80 °C to 204 °C.
Catalog No. NC3877233 Supplier Accuflow Systems Supplier No. ABW18NSF
Tygon SPT-3350 platinum-cured silicone tubing is engineered for sanitary food and beverage transfer. It features an ultra-smooth inner bore to minimize particle entrapment, low extractables to preserve fluid integrity, and a wide operating temperature range suitable for processing and sterilization.
  • Platinum-cured silicone.
  • Ultra-smooth inner bore reduces particle entrapment.
  • FDA 21 CFR 175.300 compliant.
  • NSF 51 compliant where applicable.
  • Available in sizes including 1/4 in ID x 7/16 in OD, 3/32 in wall, 50 ft coil.
  • Operating temperature range -80 °C to 204 °C.