Learn More
World Precision Instrument Saint Martin Micro Corneal Forceps, 11 cm, Black Coating
SDP
Catalog No. 502538264
Encompass
Extra-fine tip device to grab, hold, and manipulate the scleral edge of the corneal incision and other soft tissues in order to suture them during some ocular surgeries.
Catalog No. 50-253-8264 Supplier World Precision Instrument Supplier No. WPF5035
Extra-fine tip device to grab, hold, and manipulate the scleral edge of the corneal incision and other soft tissues in order to suture them during some ocular surgeries.