Learn More
World Precision Instrument Saint Martin Micro Corneal Forceps
SDP
Catalog No. 502538268
Encompass
Extra-fine tip device to grab, hold, and manipulate the scleral edge of the corneal incision and other soft tissues in order to suture them during some ocular surgeries.
Catalog No. 50-253-8268 Supplier World Precision Instrument Supplier No. WPB5035
Extra-fine tip device to grab, hold, and manipulate the scleral edge of the corneal incision and other soft tissues in order to suture them during some ocular surgeries.