Learn More
Medchemexpress LLC SAIRGA (TFA) | 99.6% | 573.64 | 1 MG
SDP
Catalog No. 5000397185
Encompass_Preferred
SAIRGA (TFA) is a chemical derived from the phages phi3T, capable of recognizing phAimR in the bacterial cytosol to induce lysogeny. This product is suitable for use as a laboratory chemical in the manufacture of various substances.
  • Solid appearance
  • Has a purity of 99.6%
  • Features a molecular weight of 573.64
  • Stable under recommended storage conditions
  • Suitable for research applications only
  • Store at -80°C for up to 2 years or -20°C for up to 1 year (powder, sealed storage, away from moisture and light)
Catalog No. 50-003-97185 Supplier Medchemexpress LLC Supplier No. HYP6012A5MG
SAIRGA (TFA) is a chemical derived from the phages phi3T, capable of recognizing phAimR in the bacterial cytosol to induce lysogeny. This product is suitable for use as a laboratory chemical in the manufacture of various substances.
  • Solid appearance
  • Has a purity of 99.6%
  • Features a molecular weight of 573.64
  • Stable under recommended storage conditions
  • Suitable for research applications only
  • Store at -80°C for up to 2 years or -20°C for up to 1 year (powder, sealed storage, away from moisture and light)