Learn More
Medchemexpress LLC Sairga (TFA) | 99.6% | 573.64 (free base) | 1 MG
SDP
Catalog No. 5000398103
Encompass_Preferred
Sairga (TFA) is a laboratory chemical primarily used for the manufacture of substances. This solid compound is stable under recommended storage conditions and is intended for research use by experienced personnel in appropriately equipped facilities.
  • Solid appearance
  • Stable under recommended storage conditions
  • Suitable for shipping at room temperature if less than 2 weeks
  • Recommended storage for powder: -80 °C for 2 years or -20 °C for 1 year
  • Recommended storage in solvent: -80 °C for 6 months or -20 °C for 1 month
  • Should be stored in a tightly sealed container in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition
Catalog No. 50-003-98103 Supplier Medchemexpress LLC Supplier No. HYP6012A1MG
Sairga (TFA) is a laboratory chemical primarily used for the manufacture of substances. This solid compound is stable under recommended storage conditions and is intended for research use by experienced personnel in appropriately equipped facilities.
  • Solid appearance
  • Stable under recommended storage conditions
  • Suitable for shipping at room temperature if less than 2 weeks
  • Recommended storage for powder: -80 °C for 2 years or -20 °C for 1 year
  • Recommended storage in solvent: -80 °C for 6 months or -20 °C for 1 month
  • Should be stored in a tightly sealed container in a cool, well-ventilated area
  • Keep away from direct sunlight and sources of ignition