Learn More
Medchemexpress LLC HY-122683 5mg Medchemexpress, sAJM589 CAS:2089-82-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1226835MG
Encompass_Preferred
Medchemexpress, HY-122683 5mg sAJM589 CAS:2089-82-9 sAJM589 is a Myc inhibitor which potently disrupts the Myc-Max heterodimer with an IC50 of 1.8 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9636