Learn More
Medchemexpress LLC HY-P1797A 5mg Medchemexpress, Sakamototide substrate peptide (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1797A5MG
Encompass_Preferred
Medchemexpress, HY-P1797A 5mg Sakamototide substrate peptide (TFA) CAS: Sakamototide substrate peptide TFA is a peptide substrate for members of the AMPK family of kinases, used in kinase activity assays [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4246