Learn More
MyBioSource SAKL ELISA KIT CANINE 96SW
SDP
Supplier:  MyBioSource MBS935908496STRIPWELLS
Encompass_Preferred
Canine Soluble Alpha-Klotho (SAKL) ELISA Kit 96-Strip-WellsMBS9359084 is a ready-to-use microwell, strip plate ELISA (enzyme-linked immunosorbent assay) Kit for analyzing the presence of the Soluble Alpha-Klotho (SAKL) ELISA Kit target analytes in biological samples. The concentration gradients of the kit standards or positive controls render a theoretical kit detection range in biological research samples containing SAKL. The ELISA analytical biochemical technique of the MBS9359084 kit is based on SAKL antibody-SAKL antigen interactions (immunosorbency) and an HRP colorimetric detection system to detect SAKL antigen targets in samples. The ELISA Kit is designed to detect native, not recombinant, SAKL. Appropriate sample types may include undiluted body fluids and/or tissue homogenates, secretions. Quality control assays assessing reproducibility identified the intra-assay CV (%) and inter-assay CV(%).For Research Use Only. Not for use in diagnostic procedures.
Catalog No. NC1943821