Learn More
IMEB, Inc Sakura Tissue Tek Slide Coverslipping Film # 4770, 1 Roll, Unique Flexible film Accepts Any Specimen, Optical Characteristics Are Comparable to Those of Glass
SDP
Supplier:  IMEB, Inc 47701
Encompass
Tissue-Tek Coverslipping Film is a unique, resin-coated plastic film that eliminates the need for cover glasses and liquid mounting media - which reduces coverslipper maintenance and enables fast coverslipping. The unique flexible film accepts any specimen. The optical characteristics are comparable to those of glass. 1 Roll (in Box).
Catalog No. 50-168-9509