Learn More
Medchemexpress LLC HY-N3006 5mg Medchemexpress, Sakuranetin CAS:2957-21-3 Purity:>98%
SDP
Catalog No. 501933935
Encompass_Preferred
Medchemexpress, HY-N3006 5mg Sakuranetin CAS:2957-21-3 Sakuranetin is a rice flavonoid phytoalexin, shows strong antifungal activity [1] . Sakuranetin has anti-inflammatory and antioxidative activities. Sakuranetin ameliorates LPS-induced acute lung injury [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3935 Supplier Medchemexpress LLC Supplier No. HYN30065MG
Medchemexpress, HY-N3006 5mg Sakuranetin CAS:2957-21-3 Sakuranetin is a rice flavonoid phytoalexin, shows strong antifungal activity [1] . Sakuranetin has anti-inflammatory and antioxidative activities. Sakuranetin ameliorates LPS-induced acute lung injury [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.