Learn More
Medchemexpress LLC Salbutamol | 18559-94-9 | MFCD00148978 | 99.5% | 1 ML
SDP
Catalog No. 5000413967
Encompass_Preferred
Salbutamol (Albuterol) is an orally active, short-acting β2-adrenergic receptor agonist, primarily used in research for bronchospasm in conditions like asthma and COPD. Studies indicate its role in gastric cancer tumorigenesis via the β2-AR/ERK/EMT pathway and its protective effect against Streptozotocin-induced skeletal muscle atrophy in diabetic rats.
  • Orally active short-acting β2-adrenergic receptor agonist.
  • Used in research for bronchospasm related to asthma and chronic obstructive pulmonary disease.
  • Induces EMT, migration, and invasion in gastric cancer cells in vitro.
  • Increases tumor growth in nude mice with gastric cancer in vivo.
  • Protects against Streptozotocin-induced skeletal muscle atrophy in diabetic rats by enhancing muscle strength, coordination, and antioxidant levels, reducing inflammation and oxidative stress, and improving muscle cellular architecture.
Catalog No. 50-004-13967 Supplier Medchemexpress LLC Supplier No. HYB103710MM/1ML
Salbutamol (Albuterol) is an orally active, short-acting β2-adrenergic receptor agonist, primarily used in research for bronchospasm in conditions like asthma and COPD. Studies indicate its role in gastric cancer tumorigenesis via the β2-AR/ERK/EMT pathway and its protective effect against Streptozotocin-induced skeletal muscle atrophy in diabetic rats.
  • Orally active short-acting β2-adrenergic receptor agonist.
  • Used in research for bronchospasm related to asthma and chronic obstructive pulmonary disease.
  • Induces EMT, migration, and invasion in gastric cancer cells in vitro.
  • Increases tumor growth in nude mice with gastric cancer in vivo.
  • Protects against Streptozotocin-induced skeletal muscle atrophy in diabetic rats by enhancing muscle strength, coordination, and antioxidant levels, reducing inflammation and oxidative stress, and improving muscle cellular architecture.