Learn More
Selleck Chemical LLC Salbutamol
SDP
Catalog No. 5000003035
Encompass_Preferred
Salbutamol (Albuterol) is a short-acting selective beta2-adrenergic receptor agonist used to treat or prevent bronchospasm in patients with asthma bronchitis emphysema and other lung diseases
Catalog No. 50-000-03035 Supplier Selleck Chemical LLC Supplier No. S549425MG
Salbutamol (Albuterol) is a short-acting selective beta2-adrenergic receptor agonist used to treat or prevent bronchospasm in patients with asthma bronchitis emphysema and other lung diseases