Learn More
Medchemexpress LLC Salbutamol (Albuterol) | 18559-94-9 | 99.5% | 250 MG
SDP
Catalog No. 5000414353
Encompass_Preferred
Salbutamol (Albuterol) is an orally active short-acting β2-adrenergic receptor agonist that promotes tumorigenesis in gastric cancer cells through the β2-AR/ERK/EMT pathway. It can relax bronchial smooth muscle, making it suitable for studying bronchospasm induced by asthma and chronic obstructive pulmonary disease.
  • Orally active short-acting β2-adrenergic receptor agonist
  • Promotes tumorigenesis in gastric cancer cells
  • Relaxes bronchial smooth muscle
  • Used to study bronchospasm in asthma and COPD
Catalog No. 50-004-14353 Supplier Medchemexpress LLC Supplier No. 250MGHYB1037
Salbutamol (Albuterol) is an orally active short-acting β2-adrenergic receptor agonist that promotes tumorigenesis in gastric cancer cells through the β2-AR/ERK/EMT pathway. It can relax bronchial smooth muscle, making it suitable for studying bronchospasm induced by asthma and chronic obstructive pulmonary disease.
  • Orally active short-acting β2-adrenergic receptor agonist
  • Promotes tumorigenesis in gastric cancer cells
  • Relaxes bronchial smooth muscle
  • Used to study bronchospasm in asthma and COPD