Learn More
Medchemexpress LLC Salbutamol-d3 | 1219798-60-3 | 98.7% | 5 MG
SDP
Catalog No. 5000413980
Encompass_Preferred
Salbutamol-d3 (Albuterol-d3) is a deuterium-labeled Salbutamol and an orally active short-acting β2-adrenergic receptor agonist. It promotes tumorigenesis in gastric cancer cells through the β2-AR/ERK/EMT pathway and can relax bronchial smooth muscle, used to study bronchospasm induced by asthma and chronic obstructive pulmonary disease.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-13980 Supplier Medchemexpress LLC Supplier No. HYB1037S5MG
Salbutamol-d3 (Albuterol-d3) is a deuterium-labeled Salbutamol and an orally active short-acting β2-adrenergic receptor agonist. It promotes tumorigenesis in gastric cancer cells through the β2-AR/ERK/EMT pathway and can relax bronchial smooth muscle, used to study bronchospasm induced by asthma and chronic obstructive pulmonary disease.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.