Learn More
Medchemexpress LLC Salbutamol-d3 (albuterol-d3) | 1219798-60-3 | MFCD06658595 | 98.6% | 242.33 g·mol⁻¹ | C13H18D3NO3 | 10 MG
SDP
Catalog No. 5000298278
Encompass_Preferred
Salbutamol-d3 is the deuterium-labeled isotopologue of salbutamol used primarily as an internal standard for quantitative analytical workflows, especially LC-MS. It is supplied as a high-purity solid in milligram-scale packages for laboratory and method development use.
  • Deuterium-labeled internal standard for accurate quantitation.
  • High chemical purity suitable for analytical methods.
  • Supplied as a solid powder for easy handling and weighing.
  • Available in multiple milligram package sizes for method development and calibration.
  • CAS number 1219798-60-3 and molecular weight 242.33 g·mol⁻¹.
  • Compatible with LC-MS workflows as an internal standard.
Catalog No. 50-002-98278 Supplier Medchemexpress LLC Supplier No. HYB1037S10MG
Salbutamol-d3 is the deuterium-labeled isotopologue of salbutamol used primarily as an internal standard for quantitative analytical workflows, especially LC-MS. It is supplied as a high-purity solid in milligram-scale packages for laboratory and method development use.
  • Deuterium-labeled internal standard for accurate quantitation.
  • High chemical purity suitable for analytical methods.
  • Supplied as a solid powder for easy handling and weighing.
  • Available in multiple milligram package sizes for method development and calibration.
  • CAS number 1219798-60-3 and molecular weight 242.33 g·mol⁻¹.
  • Compatible with LC-MS workflows as an internal standard.