Learn More
Medchemexpress LLC Salbutamol-d9 | 1173021-73-2 | 98.0% | 1 MG
SDP
Catalog No. 5000413794
Encompass_Preferred
Salbutamol-d9 (Albuterol-d9) is a deuterium-labeled short-acting β2-adrenergic receptor agonist. It serves as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. The unlabeled compound, Salbutamol, is known to promote tumorigenesis in gastric cancer cells via the β2-AR/ERK/EMT pathway and can relax bronchial smooth muscle, which is pertinent for studies concerning bronchospasm related to asthma and chronic obstructive pulmonary disease.
  • Deuterium-labeled compound for research applications.
  • Functions as a tracer for quantitative analysis.
  • Can be used as an internal standard in analytical methods.
  • Supports research in areas like asthma and chronic obstructive pulmonary disease.
  • Relevant for studies on tumorigenesis in gastric cancer cells.
Catalog No. 50-004-13794 Supplier Medchemexpress LLC Supplier No. HYB1037S21MG
Salbutamol-d9 (Albuterol-d9) is a deuterium-labeled short-acting β2-adrenergic receptor agonist. It serves as a tracer and internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. The unlabeled compound, Salbutamol, is known to promote tumorigenesis in gastric cancer cells via the β2-AR/ERK/EMT pathway and can relax bronchial smooth muscle, which is pertinent for studies concerning bronchospasm related to asthma and chronic obstructive pulmonary disease.
  • Deuterium-labeled compound for research applications.
  • Functions as a tracer for quantitative analysis.
  • Can be used as an internal standard in analytical methods.
  • Supports research in areas like asthma and chronic obstructive pulmonary disease.
  • Relevant for studies on tumorigenesis in gastric cancer cells.