Learn More
Medchemexpress LLC Salbutamol (hemisulfate) (standard) | 51022-70-9 | 99.8% | 50 MG
SDP
Catalog No. 5000408604
Encompass_Preferred
Salbutamol (Albuterol) hemisulfate (Standard) is an analytical standard intended for research and analytical applications. It is an orally active, short-acting β2-adrenergic receptor agonist. This compound promotes tumorigenesis in gastric cancer cells through the β2-AR/ERK/EMT pathway and relaxes bronchial smooth muscle, making it useful for studying bronchospasm induced by asthma and chronic obstructive pulmonary disease.
  • Analytical standard for research and analytical applications
  • Orally active short-acting β2-adrenergic receptor agonist
  • Promotes tumorigenesis in gastric cancer cells through β2-AR/ERK/EMT pathway
  • Relaxes bronchial smooth muscle
  • Used to study bronchospasm induced by asthma and chronic obstructive pulmonary disease
  • Suitable for qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-08604 Supplier Medchemexpress LLC Supplier No. HYB0436R50MG
Salbutamol (Albuterol) hemisulfate (Standard) is an analytical standard intended for research and analytical applications. It is an orally active, short-acting β2-adrenergic receptor agonist. This compound promotes tumorigenesis in gastric cancer cells through the β2-AR/ERK/EMT pathway and relaxes bronchial smooth muscle, making it useful for studying bronchospasm induced by asthma and chronic obstructive pulmonary disease.
  • Analytical standard for research and analytical applications
  • Orally active short-acting β2-adrenergic receptor agonist
  • Promotes tumorigenesis in gastric cancer cells through β2-AR/ERK/EMT pathway
  • Relaxes bronchial smooth muscle
  • Used to study bronchospasm induced by asthma and chronic obstructive pulmonary disease
  • Suitable for qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS