Learn More
Apexbio Technology LLC Salbutamol Sulfate 51022-70-9 500mg
SDP
Catalog No. 5000091384
Encompass_Preferred
Salbutamol Sulfate (CAS 51022-70-9) is a short-acting selective 2-adrenergic receptor agonist widely employed in studies related to bronchial smooth muscle relaxation The compound exerts its pharmacological effects by selectively activating 2-adrenergic receptors leading to increased intracellular cyclic AMP and subsequent airway smooth muscle relaxation Research indicates that the R-enantiomer is primarily responsible for the observed physiological effects in guinea-pig skeletal muscle with minimal contribution from the S-enantiomer Salbutamol Sulfate is commonly utilized in biomedical research focused on asthma chronic obstructive pulmonary disease (COPD) and 2-receptor pharmacodynamics
Catalog No. 50-000-91384 Supplier Apexbio Technology LLC Supplier No. B1348500
Salbutamol Sulfate (CAS 51022-70-9) is a short-acting selective 2-adrenergic receptor agonist widely employed in studies related to bronchial smooth muscle relaxation The compound exerts its pharmacological effects by selectively activating 2-adrenergic receptors leading to increased intracellular cyclic AMP and subsequent airway smooth muscle relaxation Research indicates that the R-enantiomer is primarily responsible for the observed physiological effects in guinea-pig skeletal muscle with minimal contribution from the S-enantiomer Salbutamol Sulfate is commonly utilized in biomedical research focused on asthma chronic obstructive pulmonary disease (COPD) and 2-receptor pharmacodynamics