Learn More
Cayman Chemical (R)-Salbutamol (hydrochloride), Purity: ≥98%, CAS: 50293-90-8, 50 mg
SDP
Catalog No. 502935736
Encompass_Preferred
A β2-AR agonist; induces relaxation of isolated guinea pig tracheal chains (EC50 = 16 nM); selectively binds to the β2-AR over the β1-AR (Kds = 0.236 and 1.54 µM, respectively); nebulized (R)-salbutamol reduces transpulmonary pressure in recurrent airway obstruction-affected horses (ED50 = 39.7 μg/animal)
Catalog No. 50-293-5736 Supplier Cayman Chemical Supplier No. 2399150 MG
A β2-AR agonist; induces relaxation of isolated guinea pig tracheal chains (EC50 = 16 nM); selectively binds to the β2-AR over the β1-AR (Kds = 0.236 and 1.54 µM, respectively); nebulized (R)-salbutamol reduces transpulmonary pressure in recurrent airway obstruction-affected horses (ED50 = 39.7 μg/animal)