Learn More
Medchemexpress LLC Salcaprozate-d4 sodium | 203787-91-1 | 99.7% | 305.34 | 2 MG
SDP
Catalog No. 5000435663
Encompass_Preferred
Salcaprozate-d4 (sodium) is a deuterated labeled Salcaprozate (sodium). Salcaprozate sodium (SNAC) is an oral absorption promoter and has potential as a delivery agent for oral forms of heparin and insulin. It can increase passive transcellular permeation across small intestinal epithelia due to increased lipophilicity arising from non-covalent macromolecule complexation.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Shows no toxicity to Caco-2 cells at certain concentrations.
  • Improves apparent permeability coefficient of RA and SA-B.
  • Improves oral absorption of R1 and SAs and enhances bioavailability in rats.
Catalog No. 50-004-35663 Supplier Medchemexpress LLC Supplier No. HY114299S2MG
Salcaprozate-d4 (sodium) is a deuterated labeled Salcaprozate (sodium). Salcaprozate sodium (SNAC) is an oral absorption promoter and has potential as a delivery agent for oral forms of heparin and insulin. It can increase passive transcellular permeation across small intestinal epithelia due to increased lipophilicity arising from non-covalent macromolecule complexation.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Shows no toxicity to Caco-2 cells at certain concentrations.
  • Improves apparent permeability coefficient of RA and SA-B.
  • Improves oral absorption of R1 and SAs and enhances bioavailability in rats.