Learn More
Medchemexpress LLC Salcaprozate-d4 sodium (SNAC-d4 sodium) | 203787-91-1 | 99.7% | 305.34 | 1 MG
SDP
Catalog No. 5000437320
Encompass_Preferred
Salcaprozate-d4 (sodium) is a deuterated labeled Salcaprozate (sodium). Salcaprozate sodium (SNAC) is an oral absorption promoter with potential as a delivery agent for oral forms of heparin and insulin. It can increase passive transcellular permeation across small intestinal epithelia by increasing lipophilicity from non-covalent macromolecule complexation.
  • Deuterated labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into the drug molecule.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-37320 Supplier Medchemexpress LLC Supplier No. HY114299S1MG
Salcaprozate-d4 (sodium) is a deuterated labeled Salcaprozate (sodium). Salcaprozate sodium (SNAC) is an oral absorption promoter with potential as a delivery agent for oral forms of heparin and insulin. It can increase passive transcellular permeation across small intestinal epithelia by increasing lipophilicity from non-covalent macromolecule complexation.
  • Deuterated labeled compound.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into the drug molecule.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.