Learn More
Medchemexpress LLC HY-114299 500mg , Salcaprozate (sodium) CAS:203787-91-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114299500MG
Encompass_Preferred
HY-114299 500mg Salcaprozate (sodium) CAS: 203787-91-1 Salcaprozate sodium (SNAC), an oral absorption promoter, and has the potential as a delivery agent for oral forms of heparin and insulin. Salcaprozate sodium could increase passive transcellular permeation across small intestinal epithelia based on increased lipophilicity arising from non-covalent macromolecule complexation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9317