Learn More
Cayman Chemical Salcaprozate (sodium salt), Purity: ≥98%, CAS: 203787-91-1, 500 mg
SDP
Catalog No. 502911624
Encompass_Preferred
An intestinal permeation enhancer; increases jejunal accumulation of the bioactive peptide PW5 in vitro in a non-everted rat gut sac model; increases plasma concentrations of orally administered human parathyroid hormone (1-34) in pigs
Catalog No. 50-291-1624 Supplier Cayman Chemical Supplier No. 36505500 MG
An intestinal permeation enhancer; increases jejunal accumulation of the bioactive peptide PW5 in vitro in a non-everted rat gut sac model; increases plasma concentrations of orally administered human parathyroid hormone (1-34) in pigs