Learn More
Medchemexpress LLC Salcomine | 14167-18-1 | 99.1% | 325.23 | 25 G
SDP
Catalog No. 5000453464
Encompass_Preferred
Salcomine is a compound that acts as a carrier of O2 and functions as an oxidation catalyst. It has demonstrated anti-influenza virus activity, making it suitable for research in virology and related fields.
  • Acts as a carrier of O2.
  • Functions as an oxidation catalyst.
  • Exhibits anti-influenza virus activity.
  • High purity of 99.1%.
  • Molecular weight of 325.23.
  • Identified by CAS number 14167-18-1.
Catalog No. 50-004-53464 Supplier Medchemexpress LLC Supplier No. 25GHYW013794
Salcomine is a compound that acts as a carrier of O2 and functions as an oxidation catalyst. It has demonstrated anti-influenza virus activity, making it suitable for research in virology and related fields.
  • Acts as a carrier of O2.
  • Functions as an oxidation catalyst.
  • Exhibits anti-influenza virus activity.
  • High purity of 99.1%.
  • Molecular weight of 325.23.
  • Identified by CAS number 14167-18-1.