Learn More
Active Motif Salermide
SDP
Catalog No. NC1841448
Encompass
Active Motif's Salermide is a SIRT1 and SIRT2 inhibitor. It exhibits a stronger inhibitory effect on SIRT2 than on SIRT1 in vitro. Salermide induces the reactivation of proapoptotic genes repressed by SIRT1 and causes massive apoptosis in cancer cells within 24 hours. Salermide displays protective effect in nematodes expressing muscular dystrophy protein. It promotes neuronal apoptosis confirming the protective effect of SIRT1.
Catalog No. NC1841448 Supplier Active Motif Supplier No. 14125
Active Motif's Salermide is a SIRT1 and SIRT2 inhibitor. It exhibits a stronger inhibitory effect on SIRT2 than on SIRT1 in vitro. Salermide induces the reactivation of proapoptotic genes repressed by SIRT1 and causes massive apoptosis in cancer cells within 24 hours. Salermide displays protective effect in nematodes expressing muscular dystrophy protein. It promotes neuronal apoptosis confirming the protective effect of SIRT1.