Learn More
New England Biolabs, Inc. SalI – 10000 units
SDP
Catalog No. 50812000
Encompass
A restriction endonuclease that recognizes the sequence G^TCGA_C.
Catalog No. 50-812-000 Supplier New England Biolabs, Inc. Supplier No. R0138L
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence G^TCGA_C.