Learn More
New England Biolabs, Inc. SalI – 10000 units
SDP
Catalog No. 50812001
Encompass
A restriction endonuclease that recognizes the sequence G^TCGA_C.
Catalog No. 50-812-001 Supplier New England Biolabs, Inc. Supplier No. R0138M
A restriction endonuclease that recognizes the sequence G^TCGA_C.