Learn More
New England Biolabs, Inc. SalI – 2000 units
SDP
Supplier:  New England Biolabs, Inc. R0138T
Encompass
A restriction endonuclease that recognizes the sequence G^TCGA_C.
Catalog No. 50-812-003