Learn More
New England Biolabs, Inc. SalI – 2000 units
SDP
Catalog No. 50812002
Encompass
A restriction endonuclease that recognizes the sequence G^TCGA_C.
Catalog No. 50-812-002 Supplier New England Biolabs, Inc. Supplier No. R0138S
A restriction endonuclease that recognizes the sequence G^TCGA_C.