Learn More
Apexbio Technology LLC SalI 4000U
SDP
Catalog No. 5000097066
Encompass_Preferred
SalI is a recombinant restriction endonuclease targeting the specific double-stranded DNA sequence 5 -GTCGAC-3 It is designed to recognize and cleave this DNA motif enabling DNA fragment digestion and facilitating downstream ligation reactions SalI exerts its biological activity primarily through sequence-specific endonucleolytic cleavage generating cohesive DNA ends In in vitro studies SalI demonstrates effective and rapid digestion of DNA substrates including plasmids PCR amplicons and genomic DNA Based on these properties SalI holds research potential in molecular cloning mapping of genomic regions and preparation of DNA inserts or vectors
Catalog No. 50-000-97066 Supplier Apexbio Technology LLC Supplier No. K30454000
SalI is a recombinant restriction endonuclease targeting the specific double-stranded DNA sequence 5 -GTCGAC-3 It is designed to recognize and cleave this DNA motif enabling DNA fragment digestion and facilitating downstream ligation reactions SalI exerts its biological activity primarily through sequence-specific endonucleolytic cleavage generating cohesive DNA ends In in vitro studies SalI demonstrates effective and rapid digestion of DNA substrates including plasmids PCR amplicons and genomic DNA Based on these properties SalI holds research potential in molecular cloning mapping of genomic regions and preparation of DNA inserts or vectors