Learn More
Selleck Chemical LLC Salicin 500mg 138-52-3 Salicoside,Salicine
SDP
Supplier:  Selleck Chemical LLC S2351500MG
Encompass_Preferred
Salicin (Salicoside, Salicine) is a phenol ?-glycosid produced from willow bark that shows anti-inflammatory effects. It is a natural, non-selective COX-1 and COX-2 inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-536-7