Learn More
Medchemexpress LLC Salicyl alcohol | 90-01-7 | MFCD00004617 | >99.26% | 50 G
SDP
Catalog No. 5000413382
Encompass_Preferred
Salicyl alcohol (2-Hydroxybenzyl alcohol; Saligenin) is a phenolic alcohol-type topical anesthetic and strong sensitizer with antiseptic, antibacterial and antipyretic activities. Salicyl alcohol serves as a precursor for salicin synthesis. It induces contact dermatitis and eczematous skin reactions, and is a well-known allergen identified in phenolic resins. Isolated from the barks of European aspen (Populus tremula), rowan (Sorbus aucuparia) and willow (Salix spp.), Salicyl alcohol is used in studies related to allergic contact dermatitis and is for research use only.
  • Phenolic alcohol-type topical anesthetic
  • Strong sensitizer
  • Antiseptic and antibacterial properties
  • Antipyretic activities
  • Precursor for salicin synthesis
  • Isolated from natural sources like European aspen, rowan, and willow
  • Used in studies related to allergic contact dermatitis
Catalog No. 50-004-13382 Supplier Medchemexpress LLC Supplier No. HYB141950G
Salicyl alcohol (2-Hydroxybenzyl alcohol; Saligenin) is a phenolic alcohol-type topical anesthetic and strong sensitizer with antiseptic, antibacterial and antipyretic activities. Salicyl alcohol serves as a precursor for salicin synthesis. It induces contact dermatitis and eczematous skin reactions, and is a well-known allergen identified in phenolic resins. Isolated from the barks of European aspen (Populus tremula), rowan (Sorbus aucuparia) and willow (Salix spp.), Salicyl alcohol is used in studies related to allergic contact dermatitis and is for research use only.
  • Phenolic alcohol-type topical anesthetic
  • Strong sensitizer
  • Antiseptic and antibacterial properties
  • Antipyretic activities
  • Precursor for salicin synthesis
  • Isolated from natural sources like European aspen, rowan, and willow
  • Used in studies related to allergic contact dermatitis